Polytron® Homogenizer PT 2500 E, Kinematica
Supplier: Kinematica
11010058
97057-700EA 4831.55 CAD
97057-700
Polytron® Homogenizer PT 2500 E, Kinematica
Homogenizers Benchtop Homogenizers
Kinematica Polytron® Homogenizer PT 2500 E Benchtop unit that uses the same dispersing generators as our handheld units for maximum versatility
3,5,7,12 and 20 mm generators for volumes from .05 to 2000 ml. Powerful 500 Watt motor with digital speed readout for reproducible results. 500-30,000 rpm.
Certifications: CE marked.