Orion™ AQUAfast II Chemistries Test Kits, Thermo Scientific
Supplier: Thermo Fisher Scientific
AC2002 AC2007 AC2016 AC2017 AC2027 AC2029 AC2030 AC2046 AC2048 AC2055 AC2060 AC2061 AC2065 AC2070 AC2071 AC2072 AC2078 AC2082 AC2095-WA AC2096 AC2099 AC2001 AC2012
46300-626PK 61.41 CAD
46300-626 46300-628 46300-634 46300-636 46300-640 46300-642 46300-644 46300-648 46300-650 46300-652 46300-654 46300-656 46300-658 46300-660 46300-662 46300-664 46300-668 46300-670 46300-672 46300-674 46300-676 CA46300-624 46300-632
Orion™ AQUAfast II Chemistries Test Kits, Thermo Scientific
Environmental Test Kits
Orion™ AQUAfast Tablet Reagents perform colorimetric measurements of common water parameters with convenient, rapidly dissolving tablets.
  • For use with ORION™ AQUAfast AQ4000, ORION™ AQUAfast AQ3070, ORION™ AQUAfast AQ3700, and discontinued ORION™ AQ2 series meters and other colorimeters
  • 50 to 100 tests per kit
Use Orion™ AQUAfast Tablet Reagents with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Test kits are designed for use with Orion™ AQUAfast AQ4000, Orion™ AQUAfast AQ3070, Orion™ AQUAfast AQ3700, and discontinued Orion™ AQ2 series meters and othercolorimeters.
These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging. They are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide, and zinc.