Dualmat™ Static Dissipative Mats, Bulk Roll, Royal Blue, ACL
Supplier: ACL Staticide
8285RBR2440 8285RBR3040 8285RBR3640 8285RBR4840
10856-322EA 1776.92 CAD
10856-322 10856-324 10856-326 10856-328
Dualmat™ Static Dissipative Mats, Bulk Roll, Royal Blue, ACL
Surface Protectors Bench Protectors
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer, bulk roll mats are supplied in 40 foot lengths. They are available in 24", 30", 36", and 48" widths. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes it ideal for use with constant monitors. Embossed finish reduces glare, Resistance is 10⁶ to 10⁸More Product Information