Dualmat™ Static Dissipative Mats, Precut, Green, ACL
Supplier: ACL Staticide
8185GM2436 8185GM2448 8185GM2460 8185GM2472 8185GM3060 8185GM3072 8185GM3660 8185GM3672
10856-306EA 289.01 CAD
10856-306 10856-308 10856-310 10856-312 10856-314 10856-316 10856-318 10856-320
Dualmat™ Static Dissipative Mats, Precut, Green, ACL
Surface Protectors Bench Protectors
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer precut mats feature two 10mm female snaps at the top and bottom left side. Snaps can be located elsewhere if specified. They are available in 8 standard sizes, and custom sizes are also available. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes it ideal for use with constant monitors. Embossed finish reduces glare, Resistance is 10⁶ to 10⁸ ohms. Electricals warranted for the lifetime of the mat.
Certifications: Compliant to ANSI/ESD S20.20 and RoHS.