Dualmat™ Static Dissipative Mats, Precut, Light Blue, ACL
Supplier: ACL Staticide
8085BM2448 8085BM2460 8085BM2472 8085BM3060 8085BM3660 8085BM3672
10856-282EA 68.18 CAD
10856-282 10856-284 10856-286 10856-288 10856-290 10856-292 10856-294 10856-296
Dualmat™ Static Dissipative Mats, Precut, Light Blue, ACL
Surface Protectors Bench Protectors
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer precut mats feature two 10mm female snaps at the top and bottom left side. Snaps can be located elsewhere if specified. They are available in 8 standard sizes, and custom sizes are also available. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes itMore Product Information