DuPont™ Tychem® 4000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Supplier: DuPont
SL122BWHMD001200 SL122BWHLG001200 SL122BWHXL001200 SL122BWH2X001200 SL122BWH3X001200 SL122BWH4X001200 SL122BWH2X0012BN SL122BWH3X0012BN SL122BWH4X0012BN SL122BWH5X001200 SL122BWH6X001200 SL122BWH7X001200 SL122BWHLG0012BN SL122BWHMD0012BN SL122BWHXL0012BN
CA10010-154CS 2160.43 CAD
CA10010-154 CA10010-156 CA10010-158 CA10010-160 CA10010-162 CA10010-164 10016-462 10016-464 10016-466 10016-468 10016-470 10016-472 10016-474 10016-476 10016-478
DuPont™ Tychem® 4000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. TightlyMore Product Information