HydraFlock® Dry Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
25-3316-HBT 253316HEMB80BT 25-3317-HBT 25-3318-HBT 25-3406-HBT 25-3706-HBT 25-3306-HBT
10841-238BX 647.99 CAD
10841-238 10852-166 10852-170 10852-174 10852-288 10852-292 10852-294 75788-576
HydraFlock® Dry Transport Systems, Puritan Medical Products
Swabs Transport Swabs
Get all your specimen collection devices in one place! Ideal for the collection and transport of specimen when no medium is required
Systems are designed for safe, sterile specimen transport. No matter which swab(s) are chosen, all transport systems include a round-bottom tube made from virgin polypropylene resin, which will securely hold the swab, maintaining a safe and sterile environment.
Tubes include a distinctive label describing product identity, expiry date, patient identification, and collection information. System applications include; specimen collection, rapid diagnosticMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable