PurFlock Ultra® Dry Transport Systems, Puritan Medical Products
Supplier: Puritan Medical Products
25-3406-UBT 25-3306-UBT 25-3316-UBT 25-3317-UBT 25-3318-UBT 25-3706-UBT 25-3806-UBT 25-3606-UBT
89402-812CS 925.08 CAD
89402-812 10841-236 10852-164 10852-168 10852-176 10852-290 10852-296 10852-300 10860-931
PurFlock Ultra® Dry Transport Systems, Puritan Medical Products
Swabs Transport Swabs
Get all your specimen collection devices in one place! Ideal for the collection and transport of specimen when no medium is required
Systems are designed for safe, sterile specimen transport. No matter which swab(s) are chosen, all transport systems include a round-bottom tube made from virgin polypropylene resin, which will securely hold the swab, maintaining a safe and sterile environment.
Tubes include a distinctive label describing product identity, expiry date, patient identification, and collection information. System applications include; specimen collection, rapid diagnosticMore Product Information