DuPont™ Tychem® 4000 Coveralls with Respirator Fit Hood and Attached Socks
Supplier: DuPont
SL128TWH2X000600 SL128TWH3X000600 SL128TWH4X000600 SL128TWH5X000600 SL128TWHLG000600 SL128TWHMD000600 SL128TWHSM000600 SL128TWHXL000600
10016-502CS 1149.3 CAD
10016-502 10016-504 10016-506 10016-508 10016-510 10016-512 10016-514 10016-516
DuPont™ Tychem® 4000 Coveralls with Respirator Fit Hood and Attached Socks
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Respirator fit hood
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application Cleanroom,Laboratory