Variocool Chillers, LAUDA-Brinkmann
Supplier: Lauda-Brinkmann
LWZ2104 LWZ8101 L000689 LWZ3096 LWZ3097 L000694 LWZ8095 LWZ3105 L000756 L000695 LWZ3106 LWZ8100 L000685 LWZ4095 LWZ2103 L000686 LWZ2102 L000687
10768-440EA 2455.6 CAD
10768-440 10769-038 10768-442 10769-036 10768-418 10768-410 10768-444 10768-422 10768-434 10768-446 10768-436 10768-414 10769-040 10768-448 10768-426 10768-438 10768-416 10768-428
Variocool Chillers, LAUDA-Brinkmann
Circulating Thermostats Closed Circulator Systems Cooling Circulators
LAUDA Variocool chillers offer a broad performance range for demanding temperature control tasks.
  • Color screen controller is both intuitive and informative
  • USB interface and alarm contact are standard features built into each unit
  • Option of adding additional communication interfaces for increased flexibility
LAUDA Variocool chillers were designed with a modular concept and offer a broad range of options. High powered pumps and heaters allow for a truly custom design to match your application's needs.
Certifications: CE compliant.
Ordering information: Chiller is supplied with USB and alarm contact standard. All units listed below are in 60Hz frequencies. Variant units in 50Hz configurations are also available upon request. Optional heaters and pumps are sold separately, except chiller model VC 5000 (L000756), which includes a heater (model LWZ 3096).