Cap Strips for Tubes, Molecular Bio Products
Supplier: Thermo Scientific
847-11 847-S
46300-540CS 249.31 CAD
46300-540 46300-542
Cap Strips for Tubes, Molecular Bio Products
Closures Test/Sample Tube Closures
Seal tubes tightly for storage
  • Made of polyethylene
  • For 0.75 and 1.2 mL sterile tubes
  • Available pre-sterilized as the material will not withstand autoclaving
Can be used with rows of tubes or cut apart for individual samples.