LDL-C Reagent, L-Type
Supplier: Wako Life Sciences
993-00404
999-00504
80076-150EA
1033.24
CAD
80076-150
80076-152
LDL-C Reagent, L-Type
Controls and Calibration Verification Materials
The Wako L-Type LDL-C is an in vitro assay for the quantitative determination of low density lipoprotein cholesterol (LDL-C) in serum and plasma.
See order tab for LDL-C Reagent 1 and controls.
Ordering information: Compatible with LDL-C Reagent 2 L-Type, HDL-C/LDL-C Calibrator, Control Serum I, Control Serum II, and Control Serum Combo.