cAMP and cGMP conjugates, Biotium
Supplier: Biotium
00020 00020-1 00021 00021-1 00022 00022-1
89138-100EA 511.18 CAD
89138-100 89138-102 89138-104 89138-106 89138-108 89138-110
cAMP and cGMP conjugates, Biotium
Stains Histology Stains
Conjugates for probing cAMP receptors or measuring cGMP in immunoassays.
  • Biotin-cAMP can be used to probe cAMP receptors
  • Biotin-cGMP has been used as a tracer in immunoassays for the cGMP measurement in plasma or urine samples
  • Cyanine 644-cAMP can be used to probe cAMP receptors
Cyanine 644-cAMP λEx/λEm (MeOH) = 644/670 nm
Biotin-cAMP, diisopropylethylammonium salt is a biotin-conjugated cAMP analog that can be used to probe cAMP receptors. Biotin-cGMP, diisopropylethylammonium salt has been used as a tracer in immunoassays for the cGMP measurement in plasma or urine samples. Cyanine 644-cAMP is a fluorescently labeled cAMP analog and can be used to probe cAMP receptors.
Single molecules of fluorescently labeled cAMP have been imaged as the probe binds to the receptor on the surface of Dictyostelium cells.