D-(+)-Glucose, anhydrous 97.5-102.0% USP, JP, Ph. Eur., BP suitable for use as excipient endotoxin tested

Supplier: Avantor Performance Materials

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

VWRBK876-100LB VWRBK876-12KG VWRBK876-1KG
VWRBK876-100LBEA 2345.23 CAD
VWRBK876-100LB VWRBK876-12KG VWRBK876-1KG
D-(+)-Glucose, anhydrous 97.5-102.0% USP, JP, Ph. Eur., BP suitable for use as excipient endotoxin tested
D(+)-Glucose

Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
Density: 1.54 g/cm³ (20 °C)
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity (USP) PASS
Arsenic 211 (USP) ≤ 1 μg/g
Endotoxins (USP) ≤ 2.5EU/g
Chloride 221 (USP) ≤ 0.018%
Color of Solution (USP) PASS
Dextrin (USP) PASS
Heavy Metals 231 (USP) ≤ 5μg/g
Identification A (USP) PASS
Residue on Ignition 281 (USP) ≤ 0.1%
Soluble Starch, Sulfite (USP) PASS
Specific Rotation 781S (USP) +52.6⁰ - +53.2⁰
Sulfate 221 (USP) ≤ 0.025%
Water Determination 921 (USP) ≤ 0.5%
Appearance of Solution (EP & BP) PASS
Assay (anhydrous substance) (EP & BP) 97.5% - 102.0%
Conductivity (2.2.38) (EP & BP) ≤ 20μS/cm
Dextrin (EP & BP) PASS
Identification A (Specific Optical Rotation) (EP & BP) +52.5⁰ - +53.3⁰
Identification B (EP & BP) PASS
Identification E (Water) (EP & BP) ≤ 1.0%
Related Substances (Disregard Limit) (EP & BP) ≤ 0.05%
Related Substances (Impurity C) (EP & BP) ≤ 0.2%
Related Substances (Impurity D) (EP & BP) ≤ 0.15%
Related Substances (Sum of Impurities A and B) (EP & BP) ≤ 0.4%
Related Substances (Total) (EP & BP) ≤ 0.5%
Related Substances (Unspecified Impurities) (EP & BP) ≤ 0.10%
Soluble Starch, Sulfite (EP & BP) ≤ 15ppm
Water (EP & BP) ≤ 1.0%
Acidity (JP) PASS
Arsenic 1.11 (JP) ≤ 1.3ppm
Assay (dried) (JP) ≥ 99.5%
Chloride 1.03 (JP) ≤ 0.018%
Clarity and Color of Solution (JP) PASS
Dextrin (JP) (JP) PASS
Heavy Metals (JP) ≤ 4ppm
Identification (JP) PASS
Loss on Drying 2.41 (JP) ≤ 1.0%
Residue on Ignition 2.44 (JP) ≤ 0.1
Soluble Starch, Sulfite (JP) PASS
Sulfate 1.14 (JP) ≤ 0.024%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR