Ammonium sulfate NF, ACS endotoxin tested

Supplier: Avantor Performance Materials

VWRB191-100KG VWRB191-12KG VWRB191-500G
VWRB191-100KGEA 2112.78 CAD
VWRB191-100KG VWRB191-12KG VWRB191-2.5KG VWRB191-500G
Ammonium sulfate NF, ACS endotoxin tested
Ammonium sulphate

Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.

Formula: (NH₄)₂SO₄
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
MDL Number: MFCD00003391
CAS Number: 7783-20-2

Order Now

Specification Test Results

Assay (NF) 99.0 – 105%
Iron 241 (NF) ≤ 5 ppm
Chloride 221 (NF) ≤ 5 ppm
Endotoxin (NF) ≤ 2.5000 EU/g
Residue on Ignition 281 (NF) ≤ 0.005 %
Identification (A) (NF) PASS
Identification (B) (NF) PASS
Limit of Insoluble Matter (NF) ≤ 0.005 %
Limit of Nitrate (NF) ≤ 10 ppm
Limit of Phosphate (NF) ≤ 5 ppm
pH 791 (NF) 5.0 – 6.0
Total Aerobic Microbial Count (NF) ≤1000 CFU/g
Total Combined Molds and Yeasts Count (NF) ≤ 10 CFU/g
Assay (ACS) ≥ 99.0%
Chloride (Cl) (ACS) ≤ 5 ppm
Heavy Metals (As Pb) (ACS) ≤ 5 ppm
Insoluble Matter (ACS) ≤ 0.005 %
Iron (Fe) (ACS) ≤ 5 ppm
Nitrate (NO3) (ACS) ≤ 0.001 %
pH (5% Solution at 25⁰C) (ACS) 5.0 – 6.0
Phosphate (PO4) (ACS) ≤ 5 ppm
Residue after Ignition (ACS) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR