Ward's® Preserved Grassfrogs, Single Injected
Supplier: VWR
Ward's®
WARD470001-518 WARD470000-672 WARD470001-510 WARD470000-790 WARD470000-386 WARD470000-482
470000-790PK 118.4 CAD
470000-790 470000-482 470001-518 470000-672 470000-386 470001-510
Ward's® Preserved Grassfrogs, Single Injected
Specimens Preserved Specimens Preserved Animal Materials
Formalin preserved grassfrogs. Available in vacuum packs or pails.
  • Amphibian anatomy
  • General dissection
  • High quality preservation
We offer the highest quality frog specimens for the study of body systems and general vertebrate morphology. We have plain preserved, single, and double injected grassfrogs. We carry medium (3 to 4"), large (4 to 4.5"), and extra large (4.5" and over) sizes.