Briskheat® High-Temperature Heating Tapes, Thermo Scientific
Supplier: Antyila Scientific
03105-20 03105-60 03106-60 03107-10 03118-40 03105-80 03106-20 03107-20 03107-60 03115-20 03115-40 03115-80 36119-32 36110-18
97010-140EA 151.81 CAD
97010-140 97010-148 97010-164 97010-328 97010-340 97010-348 97010-352 97010-356 97010-368 97010-384 97010-468 97010-472 97010-476 97010-484 97010-492 97010-500 97010-504 97010-152 97010-156 97010-172 97010-180 97010-236 97010-240 97010-248 97010-256 97010-264 97010-320 97010-708
Briskheat® High-Temperature Heating Tapes, Thermo Scientific
Tapes
These high-temperature heating tapes are made of resistance wires that are insulated with braided fiberglass knitted into a flat tape and covered with heavy fiberglass fabric
  • Nongrounded
  • Flexible
  • Multipurpose
  • Heavy-duty insulated design
  • Maximum exposure temperature of 1400 ºF (760 ºC)
  • Direct-contact heaters are ideal for use on conductive surfaces
Heating tapes are available with either a one-piece standard plug or a twopiece opposite-end power lead configuration. Opposite-end lead configuration provides increased flexibility when wrapping the tape around unique objects. The 120 VAC tapes include a plug and the 240 VAC tapes are supplied with bare leads. The 120 VAC tapes with an opposite-end lead configuration are supplied with a half plug on each end; the half plugs fit together and connect to form a standard 120 VAC
two-prong plug.