96-Well Flipper™ Microtube Racks, Molecular BioProducts
Supplier: Thermo Scientific
8760 8770-11
53225-610CS 790.63 CAD
53225-610 53225-612
96-Well Flipper™ Microtube Racks, Molecular BioProducts
Racks Tube Racks
Protect samples and allow racks to be stacked using this 96-well flipper microtube rack.
  • Holds 0.5 mL tubes on one side, 1.5 mL tubes on the other
  • Molded with an alphanumeric grid for easy tube identification
  • Sides have a frosted writing surface for smear-resistant labeling
  • Several colors available