Heating Tapes, Insulated, Thermo Scientific
Supplier: Antyila Scientific
36050-00 36050-10 36050-20
97010-612EA 267.62 CAD
97010-612 97010-616 97010-620 97010-624 97010-628 97010-632
Heating Tapes, Insulated, Thermo Scientific
Tapes
Dual-element design improves the heater’s flexibility over standard high-temperature tapes.
  • Greater flexibility than other high-temperature heating tapes
  • Heat to 1400 °F (760 °C)
  • Ideal for high-temperature use on conductive surfaces
Tapes are made of two heating elements that are knitted together in series and then wound together with high-temperature Samox fiberglass.