Anti-IgG2a Goat Polyclonal Antibody (CF™ 405S)

Supplier: Biotium

20381-250UL 20381-50UL
10018-282EA 440.99 CAD
10018-282 10018-284
Anti-IgG2a Goat Polyclonal Antibody (CF™ 405S)
Antibodies
Goat anti-mouse IgG2a is an affinity-purified antibody that reacts with Fc portion of the heavy chain of mouse IgG2a. To minimize cross reactivity, the antibody is cross-adsorbed against other mouse IgG subclasses (IgG1, IgG2b, IgG3), and human, bovine and rabbit serum proteins. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S results in muchMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG2a
Antigen name Immunoglobulin G2a kappa
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 405S
Reactivity Mouse
Host Goat
Isotype
Form Liquid
Purification Affinity purified
Concentration 2 mg/mL
Cross adsorption Mouse IgG1, mouse IgG2b, mouse IgG3, human, bovine and rabbit
Storage buffer PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide
Storage temperature Store at –20 °C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR