Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Supplier: Biotium

20357-200UL 20357-500UL
10018-270EA 202.52 CAD
10018-270 10018-272
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antibodies
This product is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with R-phycoerythrin (R-PE). R-PE belongs to the family of phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae. R-PE has absorption maxima at 496, 546, and 565 nm respectively, and emission at 578 nm. Because of the high extinction coefficient (~1,960,000) and high fluorescence quantum yield (~0.82), R-PE is by far the brightest dye for the 565 nm wavelength region.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR