Anti-IgA Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20427-500UL 20427-50UL
10018-344EA 342.99 CAD
10018-344 10018-346
Anti-IgA Goat Polyclonal Antibody (CF™ 633)
Antibodies
Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™633 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™633 (Ex/Em 630/650 nm) produces brighter antibody conjugates than Alexa Fluor® 647, and has unmatched photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.





Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: CF633
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR