Anti-IgA Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20428-500UL 20428-50UL
10018-350EA 342.99 CAD
10018-350 10018-352
Anti-IgA Goat Polyclonal Antibody (CF™ 488A)
Antibodies
Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™488A dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Green fluorescent CF™488A (Ex/Em 490/515 nm) is a superior alternative to FITC (or fluorescein) owing to its exceptional brightness and photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR