Anti-IgG Lama Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20458-500UL 20458-50UL
10001-942EA 360.95 CAD
10001-942 10001-944
Anti-IgG Lama Polyclonal Antibody (CF™ 647)
Antibodies
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Mouse IgG
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 647
Reactivity Mouse
Host Llama
Isotype
Form Liquid
Purification Affinity purified
Concentration 2 mg/mL
Cross adsorption No
Storage buffer PBS, 50% glycerol, 2 mg/ml BSA, 0.05% sodium azide
Storage temperature Store at –20 °C.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR