Anti-IgG Lama Polyclonal Antibody (CF™ 488A)
Supplier: Biotium
20454-500UL 20454-50UL
10001-936EA 360.95 CAD
10001-936 10001-938
Anti-IgG Lama Polyclonal Antibody (CF™ 488A)
Antibodies
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™488A (Ex/Em 490/515 nm) is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF™488A has brightness and photostability comparable to Alexa Fluor® 488.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
More Product Information