Anti-IgG Lama Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20452-500UL 20452-50UL
10001-924EA 360.95 CAD
10001-924 10001-926
Anti-IgG Lama Polyclonal Antibody (CF™ 640R)
Antibodies
Llama anti-rabbit CF™ dye conjugates are prepared by labeling affinity purified llama anti-rabbit IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Llama
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR