Anti-IgG gamma Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20447-500UL 20447-50UL
10001-894EA 421.39 CAD
10001-894 10001-896
Anti-IgG gamma Goat Polyclonal Antibody (CF™ 640R)
Antibodies
Goat anti-human IgG, Fc gamma specific antibody reacts specifically with the human IgG heavy (γ) chains and not with light chains and F(ab’)2 fragment on all human immunoglobulins. To minimize cross-reactivity, it has been adsorbed against bovine, horse, mouse and rabbit serum proteins prior to conjugation. CF™640R (Ex/Em 642/662 nm) is a novel far-red rhodamine-based dye spectrally similar to Cy®5 and Alexa Fluor® 647. CF™640R is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647 when excited at 633 or 635 nm. However, the most important advantage of CF™640R is its exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR