Metal Hub Needles, Hamilton
Supplier: Hamilton
91020 91021 91024 91025
10217-422PK 150.14 CAD
10217-422 10217-424 10217-426 10217-428
Metal Hub Needles, Hamilton
Syringe Needles
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors.
  • Autoclavable
  • 304 stainless steel needles
  • Custom needle lengths and point styles are available
Packaging: Conveniently packed with 6 needles per pack.