SpecMat-H Anti-Static Mats, Precut, 2.54 mm Thickness, ACL
Supplier: ACL Staticide
6212436 6212448 6212460 6212472 6233060 6233072 6253660 6253672
10856-236EA 224.68 CAD
10856-236 10856-238 10856-240 10856-242 10856-244 10856-246 10856-248 10856-250
SpecMat-H Anti-Static Mats, Precut, 2.54 mm Thickness, ACL
Surface Protectors Bench Protectors
SpecMat-H Mats are static dissipative, single-layer, ESD-safe thermoplastic vinyl mats made for bench tops and other work surfaces
These single layer mats include two 10mm female snaps at top and bottom left side. Snaps can be located elsewhere if specified.
Available in 8 standard sizes. Custom sizes are also available. Mats are 100% recyclable, and made in the USA.
They also work as an excellent shelving liner. Lifetime warranty on electrical resistance (10⁸ to 10⁹ ohms).
Certifications: Compliant to ANSI/ESDA S20.20.