SpecMat-H Anti-Static Mats, Bulk Rolls, 2.54 mm Thickness, ACL
Supplier: ACL Staticide
62100 62300 62500
10856-230EA 1713.98 CAD
10856-230 10856-232 10856-234
SpecMat-H Anti-Static Mats, Bulk Rolls, 2.54 mm Thickness, ACL
Surface Protectors Bench Protectors
SpecMat-H Mats are static dissipative, single-layer, ESD-safe thermoplastic vinyl mats made for bench tops and other work surfaces
Mats are 100% recyclable, and made in the USA. They are available in 24", 30", and 36" widths. All bulk roll mats are 50' in length.
They also work as an excellent shelving liner. Lifetime warranty on electrical resistance (10⁸ to 10⁹ ohms).
Certifications: Compliant to ANSI/ESDA S20.20.