Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Supplier: Biotium
10003 10004 10005 10006 10007 10012 10013 10014 10015 10017 10018 10019 10020 10022 10024 10025 10030
89138-192EA 272.49 CAD
89138-192 89138-194 89138-196 89138-198 89138-200 89138-222 89138-224 89138-226 89138-228 89138-232 89138-234 89138-236 89138-238 89138-240 89138-242 89138-244 89138-248
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Protein Linking and Labeling Systems
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
  • Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
  • Chromogenic substrates for absorbance-based detection or light microscopy
  • Fluorogenic substrates for highly sensitive fluorescence-based ELISA
  • Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.