VWR® Glassware Washers with DI Water Booster Pump
Supplier: SP Industries
100004796 10002-872 10002-874 10002-876
10002-870EA 17837.11 CAD
10002-870 10002-872 10002-874 10002-876
VWR® Glassware Washers with DI Water Booster Pump
Glassware Washers
Designed for simple operation and flexible loading, Glassware Washers with DI Rinse and external DI Water Booster Pump can accommodate a wide range of requirements. The included external DI water pump is recommended when the incoming DI water source has a pressure of less than 18psi. Booster pump packaged separately for installation on site.
  • 304 stainless steel washing chambers ensure long life and corrosion protection; 304SS exterior door for Undercounter models; 304SS full exterior casing for Freestanding/Mobile units
  • Enhanced dual fan drying system
  • Temperature selection to 70 °C (158 °F) for optimal cleaning of different loads
  • Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy cost
  • Spindle Rack Ready models can accommodate 1 or 2 direct injection spindle racks
The integrated DI water pump is recommended when the incoming DI water source has a pressure of less than 18psi.
Three separate filters protect pumps from debris and are conveniently located for easy access. Multiple cycle selections and microprocessor control systems ensure that each load is processed efficiently with minimum energy consumption.
New improved units feature a soft touch controller, improved drying, a quieter run, and are compatible with existing racks and accessories. Engineered for reliable and energy efficient operation, these durable washers are available in four models: Undercounter Washer with DI Rinse and DI Pump; with or without Spindle Rack Ready, and Freestanding/Mobile Washer with DI Rinse and DI Pump; with or without Spindle Rack Ready.
Certifications: ETL and ETL-C listed.
Ordering information: Freestanding/Mobile models supplied with mounting feet and wheels. A wide range of accessories, including spindle racks, specialty baskets, and inserts accommodating specific laboratory glassware are available to fit both upper and lower racks. For more information, contact your Avantor representative.
Delivery information: All models come with top and bottom stainless steel open racks to accommodate a variety of glassware; spindle racks must be purchased separately.
Caution: Notwithstanding any other Avantor terms and conditions of sale, an order of this product may not be cancelled and the product may not be returned except due to product defects or receipt of an incorrect product.