Reactive Biotin Compounds, Biotium
Supplier: Biotium
90050 90051 90052 90053 90054 90058 90059 90061 90063 90064 90064-1 90065 90065-1 90066 90066-1 90067 90068 90069 90078 90079 90080 90083 90111 90112 90113 92010 92011 92012 92224 92176 92167 92168 92169
89139-600EA 155.03 CAD
89139-600 89139-602 89139-604 89139-606 89139-608 89139-616 89139-618 89139-622 89139-626 89139-628 89139-630 89139-632 89139-634 89139-636 89139-638 89139-640 89139-642 89139-644 89139-658 89139-660 89139-662 89139-668 89139-722 89139-724 89139-726 89139-800 89139-802 89139-804 89411-598 10119-154 10119-220 10119-222 10119-224
Reactive Biotin Compounds, Biotium
Protein Expression Protein Expression Reagents
Biotinylation reagents with a variety of reactive functional groups.
  • Reagents for biotinylation of proteins and other biomolecules
  • Full selection of reactive functional groups
  • Variety of linker lengths available for improved water solubility and streptavidin binding
Biotium offers reactive biotin compounds with a variety of functional groups:
Amine reactive:
Succinimidyl esters react with primary and secondary amines and can be used to label proteins or peptides
The Biotin SE Protein Labeling Kit includes everything you need to label and purify three reactions of up to 1 mg protein each with biotin SE
Thiol reactive:
Maleimide and iodoacetamide react with thiol groups
MTSEA selectively labels thiols under mild conditions
TS biotin compounds allow selective labeling of cell surface thiols
Other reactive chemistries:
Amine groups can be reacted with activated carboxylic acids
Cadaverine can be coupled to carboxylic acids, DNA, and other biomolecules
Cadaverine free base can be directly coupled to activated carboxylic acids without the use of additional base
Hydrazides react with aldehydes or ketones to form unstable hydrazone linkages
Aminooxy reacts with aldehydes or ketones to form stable oxime bonds, and may be preferable to hydrazides for labeling
Tyramides can be used for HRP-mediated tyramide signal amplification (TSA) reactions
Alkynes react with azides by copper(I)-catalyzed 1,3-dipolar Huisgen cycloaddition
BCN groups react with azides by copper-free 1,3-dipolar Huisgen cycloaddition
Also see our selection of fluorescent CF™ Dye Streptavidin and Monoclonal Mouse Anti-Biotin Conjugates.