Kel-F® Hub Needles, Hamilton
Supplier: Hamilton
90521 90536 90514 90516 90517 90535 90520 90534 90523 90524 90525 90533 90539 90532 90528 90530 90531
30618-692PK 216.24 CAD
30618-692 10217-442 10217-456 10217-460 10217-462 10217-464 10217-468 10217-470 10217-472 10217-474 10217-476 10217-478 10217-480 10217-482 10217-484 10217-488 10217-490
Kel-F® Hub Needles, Hamilton
Syringe Needles
Kel-F® hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors.
  • Autoclavable, gas sterilizable
  • 304 stainless steel needles
Packaging: Conveniently packed with 6 needles per pack.