CM2800 Series Dual Wire Resistance Monitor, Transforming Technologies
Supplier: Transforming Technologies
CM2800 CM2815
10837-018EA 1119.19 CAD
10837-018 10837-020
CM2800 Series Dual Wire Resistance Monitor, Transforming Technologies
Static Control Monitors
The CM2800 Series Dual Conductor workstation monitor provides complete ESD protection by grounding and providing continuous, uninterrupted monitoring of most grounding parameters of operators, workbenches, and tools
The CM2800 alarm settings can be reconfigured quickly and easily with the PC software CM2800-SEE. Operator Resistance, Operator Voltage, Work-surface Resistance, Operator Presence Check (OPC), and Resistance of Equipment alarms can be enabled/disabled and limits selectable to user specifications. The alarm statuses are displayed using tricolor LED’s on the front panel of theMore Product Information