VWR® Straight Sided Jars, Clear and Amber
Supplier: VWR International
10861-932 10861-934 10861-940 10861-942 10861-962 10861-964 10861-966 10861-968 10861-972 10861-992 10861-994 10861-996 10862-028 10862-030 10862-036 10862-038 10862-046 10862-048 10862-054 10862-056 10862-060 10862-068 10862-070 10862-074 10862-076 10862-078 10862-080 10862-112 10862-122 10862-124 10862-128 76329-550 76329-560 76329-566 76329-572 76329-628 10862-059 77394-648
10861-932CS 255.17 CAD
10861-932 10861-934 10861-938 10861-940 10861-942 10861-962 10861-964 10861-966 10861-968 10861-972 10861-992 10861-994 10861-996 10862-024 10862-028 10862-030 10862-032 10862-036 10862-038 10862-044 10862-046 10862-048 10862-054 10862-056 10862-060 10862-068 10862-070 10862-074 10862-076 10862-078 10862-080 10862-112 10862-120 10862-122 10862-124 10862-128 76329-550 76329-560 76329-566 76329-572 76329-628 10862-059 77394-648
VWR® Straight Sided Jars, Clear and Amber
Jars General Use Jars
VWR® Straight Sided Jars are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • Clear and amber options
  • Straight sides, without shoulders for maximum storage capacity
Jars are available in multiple packaging options.
Order Now
SPECIFICATIONS
Environmentally Preferable