VWR® Bottles, Clear, Wide Mouth
Supplier: VWR International
293155 VWR89043-842 295147
10862-344CS 190.89 CAD
10862-344 10862-346 10862-348 10862-352 10862-354 76359-572
VWR® Bottles, Clear, Wide Mouth
Bottles Glass Bottles
VWR® Clear wide mouth bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
Two packaging options are available for these wide mouth bottles. They may be purchased in convenience packs with closures attached to help maintain cleanliness, or in bulk packs with unattached closures included separately in bags.