VWR® French Square Bottles, Clear
Supplier: DWK Life Sciences (KIMBLE)
10862-086 VWR89043-670 10862-094 10862-096 10862-098 10862-100 10862-102 10862-106 VWR89043-734 VWR89043-738 VWR89043-740 10862-144 10862-146 10862-148 10862-150 10862-154 10862-156 10862-180 10862-182 10862-184 10862-188 10862-192 10862-194 10862-198 10862-200 10862-202 10862-204 10862-206 10862-342 76404-282
10862-082CS 1091.43 CAD
10862-082 10862-086 10862-088 10862-094 10862-096 10862-098 10862-100 10862-102 10862-106 10862-108 10862-110 10862-136 10862-138 10862-140 10862-144 10862-146 10862-148 10862-150 10862-154 10862-156 10862-180 10862-182 10862-184 10862-188 10862-190 10862-192 10862-194 10862-198 10862-200 10862-202 10862-204 10862-206 10862-342 76404-282
VWR® French Square Bottles, Clear
Bottles Glass Bottles
VWR® clear french square style bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
Three packaging options are available for these VWR® french square bottles. They may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures included in separate bags, or bulk packed without closures.
Packaging: Available in a variety of package sizes with different closure. options