VWR® Tablet Bottles, Clear, Glass, Square
Supplier: DWK Life Sciences (KIMBLE)
VWR89044-464 VWR89044-468
10862-384CS 1580.4 CAD
10862-384 10862-386
VWR® Tablet Bottles, Clear, Glass, Square
Bottles Glass Bottles
VWR® Clear glass square tablet bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • Available in a variety of package sizes with different closure options
Two packaging options are available for these bottles. They may be purchased in bulk packs with unattached closures included in separate bags, or bulk packed without closures.
Order Now
SPECIFICATIONS
Environmentally Preferable