Bovine Serum Albumin (BSA), Fraction V
Supplier: MilliporeSigma
126609-100GM 126609-10GM 126609-5GM
CA80055-702EA 3312.2 CAD
CA80055-702 CA80055-704 CA80055-706
Bovine Serum Albumin (BSA), Fraction V
Proteins and Peptides
Albumin, Bovine Sera, is designed for use in cell culture systems and animal perfusion studies.
  • Nuclease-free
  • Protease-free