SINGLE StEP® Empty Column, Thomson Instrument Company
Supplier: THOMSON INSTRUMENT CO
9452092-100 94520-10 9452088-100
76808-368CS 1936.23 CAD
76808-368 76808-364 76808-380
SINGLE StEP® Empty Column, Thomson Instrument Company
Chromatography Columns
Thomson Instrument Company is aware of the need to customize an available apparatus to individual experiments
The SINGLE StEP® Empty Columns allow users the opportunity to fill different sized columns with resins for purification applications. The wide range of columns offered means the chemist does not have be limited by column sizes. Empty columns also allow for the ability to connect these columns to FPLC systems.