VWR® Phenolic Resin Worksurfaces for Carts, Black
Supplier: VWR
AMCTWB3572H AMCTWB3172M AMCTWB3548M AMCTWB3160M AMCTWB3136M AMCTWB3130H AMCTWB3530M AMCTWB3160H AMCTWB3172H AMCTWB3548H AMCTWB3572M AMCTWB3148H AMCTWB3536H AMCTWB3130M AMCTWB3560H AMCTWB3536M AMCTWB3530H AMCTWB3136H AMCTWB3560M AMCTWB3148M
AMCTWB3536HEA 2618.53 CAD
AMCTWB3536H AMCTWB3548H AMCTWB3548M AMCTWB3136H AMCTWB3148H AMCTWB3172H AMCTWB3560M AMCTWB3572M AMCTWB3130H AMCTWB3136M AMCTWB3148M AMCTWB3530H AMCTWB3536M AMCTWB3572H AMCTWB3160M AMCTWB3172M AMCTWB3130M AMCTWB3560H AMCTWB3530M AMCTWB3160H
VWR® Phenolic Resin Worksurfaces for Carts, Black
Furniture Tables and Workbenches
Phenolic resin worksurfaces are composed of cellulose fiber-reinforced, phenolic resin core with a highly crosslinked, polyurethane, copolymer surface.
  • Each worksurface has three square edges and one edge with a ¹/₈" chamfer
  • For High-Column worksurfaces, two or three cut-outs accomodate 76" high columns, depending on bench width
  • High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Phenolic resin worksurfaces are chemical-resistant and non-flammable.