VWR® Plastic Laminate Worksurface
Supplier: VWR
AMCLWB3536H AMCLWB3130M AMCLWB3548H AMCLWB3530M AMCLWB3572H AMCLWB3536M AMCLWB3572M AMCLWB3560M AMCLWB3530H AMCLWB3160H AMCLWB3136M AMCLWB3172H AMCLWB3148H AMCLWB3136H AMCLWB3130H AMCLWB3160M AMCLWB3148M AMCLWB3548M AMCLWB3172M AMCLWB3560H
AMCLWB3572MEA 1236.3 CAD
AMCLWB3572M AMCLWB3560M AMCLWB3172H AMCLWB3136M AMCLWB3130H AMCLWB3148M AMCLWB3148H AMCLWB3136H AMCLWB3548H AMCLWB3536H AMCLWB3548M AMCLWB3536M AMCLWB3530H AMCLWB3172M AMCLWB3572H AMCLWB3160M AMCLWB3130M AMCLWB3560H AMCLWB3530M AMCLWB3160H
VWR® Plastic Laminate Worksurface
Furniture Tables and Workbenches
Laminate worksurfaces are composed of high-pressure plastic laminate.
  • For High-Column worksurface two or three cut-outs accomodate 76" high columns, depending on bench width
  • High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Each worksurface has three square edges and one full 180-degree wrapped front edge.