Dehydrant Blends, Epredia
Supplier: RICHARD-ALLAN SCIENTIFIC CA
6201 6501 6301 6355 6401
CA66004-946CS 449.6 CAD
CA66004-946 CA66004-948 CA84000-046 CA84000-048 CA84000-050
Dehydrant Blends, Epredia
Stains Microscopy and staining reagents
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.