Difco™ Noble Agar, BD Biosciences
Supplier: BD Biosciences
214230 214230
CA90000-774EA 1425.77 CAD
CA90000-774 90000-774
Difco™ Noble Agar, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Noble Agar is suitable for immunodiffusion, some electrophoretic applications and as a substrate for mammalian or plant tissue culture.
  • Suitable for immunodiffusion
  • Substrate for mammalian or plant tissue culture
  • Extensively washed and bleached
Extensively washed and bleached. This agar should be used for applications where extreme clarity and high purity are required.