VWR® Kemresin® Epoxy Resin Worksurfaces for Carts
Supplier: VWR International
AMCRWB3130H AMCRWB3130M AMCRWB3136H AMCRWB3136M AMCRWB3148H AMCRWB3148M AMCRWB3160H AMCRWB3160M AMCRWB3172H AMCRWB3172M AMCRWB3530H AMCRWB3530M AMCRWB3536H AMCRWB3536M AMCRWB3548H AMCRWB3548M AMCRWB3560H AMCRWB3560M AMCRWB3572H AMCRWB3572M
AMCRWB3130HEA 1638.6 CAD
AMCRWB3130H AMCRWB3130M AMCRWB3136H AMCRWB3136M AMCRWB3148H AMCRWB3148M AMCRWB3160H AMCRWB3160M AMCRWB3172H AMCRWB3172M AMCRWB3530H AMCRWB3530M AMCRWB3536H AMCRWB3536M AMCRWB3548H AMCRWB3548M AMCRWB3560H AMCRWB3560M AMCRWB3572H AMCRWB3572M
VWR® Kemresin® Epoxy Resin Worksurfaces for Carts
Furniture Tables and Workbenches
These work surfaces are composed of molded, modified epoxy resin.
  • Epoxy resin fabrication
  • Worksurfaces can be used at different heights
Each worksurface has three square edges and one edge with a ¹/₁₈" chamber. For High Column worksurface two or three cut-outs accomodate 76" high columns, depending on bench width. High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required.