eXtreme/FV® Filter Vial, Thomson Instrument Company
Supplier: THOMSON INSTRUMENT CO
85530-500 85531-200 85531-500 85535-200 85535-500 85539-200 85539-500 85540-200 85540-500 85541-200 85541-500 85530-200
76808-666CS 3199.18 CAD
76808-666 76808-654 76808-668 76808-656 76808-670 76808-658 76808-672 76808-660 76808-674 76808-662 76808-676 76808-664 76808-678 76808-652
eXtreme/FV® Filter Vial, Thomson Instrument Company
Filters Syringeless Filters
Thomson's filtration products are engineered and optimized to increase the efficiency of sample preparation without compromising the results.
Thomson eXtreme/FV® Filter Vials offer tiered filtration, and are designed to speed up sample prep and analysis on particulate laden samples. Save on costly repairs and service issues while eliminating multiple syringe filters, syringes, HPLC vials, and HPLC caps. The eXtreme/FV® Filter Vials are easy to press down with hands or toggle press.
These filter vials are manufactured with attention to detail and high-grade quality to ensure performanceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable