nano|Filter Vials, Thomson Instrument Company

Supplier: THOMSON INSTRUMENT CO
25535-200 25535-500 25538-200 25538-500 25540-200 25540-500 25541-200 25541-500 15530-200 15530-500 15531-200 15531-500 15535-200 15535-500 15538-200 15538-500 15540-200 15540-500 15541-200 15541-500 25530-200 25530-500 25531-200 25531-500 14638
76808-590CS 1317.91 CAD
76808-590 76808-592 76808-594 76808-596 76808-598 76808-600 76808-602 76808-604 76808-558 76808-560 76808-562 76808-564 76808-566 76808-568 76808-570 76808-572 76808-574 76808-576 76808-578 76808-580 76808-582 76808-584 76808-586 76808-588 77497-284
nano|Filter Vials, Thomson Instrument Company
Filters Syringeless Filters
Thomson nano|Filter Vials offer a low dead volume, allowing filtration with as little as 10 µl of sample with enough remaining filtrate to make a 2 µl injection.

  • In vial filtration with minimal sample loss, 10 µl sample for a 2 µl injection
  • Minimize sample loss by eliminating multiple transfers
  • Compatible with standard UHPLC or HPLC autosamplers (12×32 mm)
  • Available in 0.2 µm and 0.45 µm pore sizes
  • Split and non-split septums available

Minimal sample loss makes the nano|Filter Vials ideal for critical samples replacing syringe filters, spin columns, autosampler vial, low volume insert, and cap.

Simply add the sample and sample buffer, press, and it is ready for the UPLC or HPLC.

Accessories information: The accessory Filter Vial Presses quickly depress autosampler-ready filter vials.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR