Mini Fan, Nederman
Supplier: Nederman
70332961 70332961
CA89087-698EA 1546.48 CAD
VF332961 CA89087-698
Mini Fan, Nederman
Fume Extractors
The Mini Fan offers a 175cfm capacity, with 12.7cm (5") inlet and outlet ports
This fan is not equipped with a speed controller.