Grippy® Surgical Absorbent Mat Roll, PIG®
Supplier: New Pig
HC101 HC102 HC103 HC100
10031-708RL 348.39 CAD
10031-708 10031-710 10031-712 10031-714
Grippy® Surgical Absorbent Mat Roll, PIG®
Spill Control Materials
Mat roll with easy tear perforations absorbs saline and blood and stays put to keep O.R. floors dry and safe during high fluid surgeries.
  • Mat roll sticks to the floor and won't slide, shift or bunch up during procedures; wide heavyweight rolls are perfect for absorbing large amounts of fluids around O.R. tables and lab stations
  • Absorbs up to 3 O.R. fluids including blood, vomit, urine, saline and water
  • More absorbent, more sanitary, faster and easier to use than towels, paper towels and other O.R. clean-up methods
  • Disposable mats keep the surgical
More Product Information